Four-Piece Single-End Probe Set, Aven Tools
# de catalogue 47743-166
Fournisseur: Aven Tools
Spécifications
- Description:Four-Piece Single-End Probe Set
- Cat. no.:47743-166
- Supplier No.:20210
Spécifications
A propos de cet article
Scalpel-shaped for fine probing, these tools are useful for cold solder probing, hand inspection, micro positioning, lead adjusting, and more
Made of stainless steel.
Customers Who Bought This Also Bought