Nalgene® Filter Storage Bottle Receivers, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These receiver storage bottles feature an ergonomic design with tapered sides and grips for ease of handling.
- Integral no-drip lip facilitates pouring
- Easy-to-read graduations on two sides
- Non-cytotoxic, non-pyrogenic, radiation sterilized
Storage bottles have a 45 mm Ø closure and are designed for use with Nalgene® Rapid-Flow™ series bottle-top filters and filter units. A 1¹/₂ -turn threaded screw closure guarantees a leakproof seal and reduces pH shift of stored media. The storage bottles have at least 20% extra head space for the addition of serum or other additives to the filtrate.
Specifications
- Catalog No:
- 28199-225
- 28199-214
- 28199-246
- 28199-189
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)