CuffStrips™, Sleeve Seal Tape, Micronova
Supplier: Micronova
Certificates
About this item
Conformable pre-cut polyethylene tape strips are used to ensure an effective seal between the glove and garment in critical environments.
- Medium /Low-tack available for applications needing firmer hold in the Dark Blue version
- Irradiated strips are suitable for aseptic processing and production and food applications
- Available in 2" and 3" widths
- Available in flat configuration and trifold configuration for more compact storage
Low-tack acrylic adhesive removes cleanly and leaves no adhesive residue or damage to garments.
Packaging: Packaged as 45.7 cm (18") pre-cut strips bagged in pairs, 400 each, 200 pairs per case.
Customers Who Bought This Also Bought