KIMBLE® Microscale Threaded Chemistry Kits, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
CE Compliant
Certificats
A propos de cet article
These threaded chemistry kits contain some of the components necessary to perform the experiments described in the following texts: organic laboratory techniques: microscale approach, Pavia, Lampman, Kriz, Engel, Saunders college publishing and microscale organic laboratory, Mayo, Pike, Butcher, John Wiley and Sons.
- The rugged polyethylene storage case (2.5H×13.5W×10"D) features a die cut foam insert
- The EKONO-CASE™ is a low-cost corrugated cardboard alternative to plastic
- Kits fit easily into lab bench drawers
- Standard taper 14/10 connections
- High temperature-resistant blue nylon connectors
- Ideal for use up to 200 °C
Kit components are manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirement.
Spécifications
- Catalog No:
- KT746210-0005
- KT746000-0005
- KT746210-0000
- KT746000-0000
- KT746220-0000
- KT746200-0005
- KT746200-0000
- Description:
- Threaded starter kit in EKONO-CASE™
- Threaded standard kit in EKONO-CASE™
- Threaded starter kit in polyethylene storage case
- Threaded standard kit in polyethylene storage case
- Threaded basic kit in polyethylene storage case
- Threaded intermediate kit in EKONO-CASE™
- Threaded intermediate kit in polyethylene storage case
- Kit contents:
- 1 Claisen adapter
1 jacketed condenser
1 Hickman distillation head
1 Craig recrystallization tube, 2 ml
1 Craig recrystallization plug
1 drying tube
1 pear shaped flask, 5 ml, thin-wall
1 micro vial, 5 ml
6 compression cap, 20-400
4 O-ring, ethylene propylene, size 112
2 septum
1 syringe, polyethylene
1 gas delivery tube
1 centrifuge tube, 15 ml
1 foam insert set
1 storage case - 1 Claisen adapter
1 connecting adapter
1 thermometer adapter
1 jacketed condenser
1 air condenser
1 Hickman distillation head
1 Craig recrystallization tube, 2 ml
1 Craig recrystallization plug
1 drying tube
1 pear shaped flask, 5 ml, thin-wall
1 round bottom flask, 10 ml
1 micro vial, 0.1 ml
1 micro vial, 3 ml
1 micro vial, 5 ml
1 GC collection tube with O-ring
1 spin vane
12 compression cap, 20-400
8 O-ring, E.P., size 112
4 septum
1 syringe, polyethylene
1 gas delivery tube
1 centrifuge tube, 15 ml
1 vacuum adapter
1 foam insert set
1 storage case, black PE - 1 Claisen adapter
1 jacketed condenser
1 Hickman distillation head
1 Craig recrystallization tube, 2 ml
1 Craig recrystallization plug
1 drying tube
1 pear shaped flask, 5 ml, thin-wall
1 micro vial, 5 ml
6 compression cap, 20-400
4 O-ring, ethylene propylene, size 112
2 septum
1 syringe, polyethylene
1 gas delivery tube
1 centrifuge tube, 15 ml
1 foam insert set
1 storage case - 1 Claisen adapter
1 connecting adapter
1 thermometer adapter
1 jacketed condenser
1 air condenser
1 Hickman distillation head
1 Craig recrystallization tube, 2 ml
1 Craig recrystallization plug
1 drying tube
1 pear shaped flask, 5 ml, thin-wall
1 round bottom flask, 10 ml
1 micro vial, 0.1 ml
1 micro vial, 3 ml
1 micro vial, 5 ml
1 GC collection tube with O-ring
1 spin vane
12 compression cap, 20-400
8 O-ring, E.P., size 112
4 septum
1 syringe, polyethylene
1 gas delivery tube
1 centrifuge tube, 15 ml
1 vacuum adapter
1 foam insert set
1 storage case, black PE - 1 Claisen adapter
1 jacketed condenser
1 Hickman distillation head
1 Craig recrystallization tube, 2 ml
1 Craig recrystallization plug, PTFE
1 drying tube
1 micro vial, 3 ml
1 micro vial, 5 ml
7 compression cap, 20-400
5 O-ring, E.P., size 112
2 septum
1 round bottom flask, 10 ml
1 universal inlet/vacuum adapter
1 foam insert set
1 storage case - 1 Claisen adapter
1 thermometer adapter
1 jacketed condenser
1 air condenser
1 Hickman distillation head
1 vacuum adapter
1 Craig recrystallization tube, 2 ml
1 Craig recrystallization plug
1 drying tube
1 pear shaped flask, 5 ml, thin-wall
1 round bottom flask, 10 ml
1 micro vial, 3 ml
1 micro vial, 5 ml
10 compression cap, 20-400
6 O-ring, E.P., size 112
4 septum
1 syringe, polyethylene
1 gas delivery tube
1 centrifuge tube, 15 ml
1 foam insert set
1 storage case - 1 Claisen adapter
1 thermometer adapter
1 jacketed condenser
1 air condenser
1 Hickman distillation head
1 vacuum adapter
1 Craig recrystallization tube, 2 ml
1 Craig recrystallization plug
1 drying tube
1 pear shaped flask, 5 ml, thin-wall
1 round bottom flask, 10 ml
1 micro vial, 3 ml
1 micro vial, 5 ml
10 compression cap, 20-400
6 O-ring, E.P., size 112
4 septum
1 syringe, polyethylene
1 gas delivery tube
1 centrifuge tube, 15 ml
1 foam insert set
1 storage case