Cleanroom Tape Dispensers, Ultratape
Fournisseur: Ultratape Industries
Certificats
A propos de cet article
Dispensers feature heavy, all-metal construction with a 7.6cm (3") plastic core, formed steel frame and a nonslip rubber facing on the bottom
Single roll dispenser 11212-142 accepts one roll of tape up to 5.1cm (2") wide x 201m (220yds.) long. Multiple roll dispenser 11212-144 accepts four rolls of tape up to 2.5cm (1") wide x 201m (220yds.) long.
Serrated blade cuts all non-reinforced tapes quickly and cleanly and can be replaced. Unit is powder-coated with an epoxy-polyester finish. Two holes on the dispenser bottom allow permanent mounting to benchtop or ESD grounding.
Customers Who Bought This Also Bought