SP Wilmad-LabGlass Microchemistry Joint Kits, SP Industries
Fournisseur: Avantor
Certificats
A propos de cet article
Choose kits with either [ST]14/20 or [ST]19/22 joints
Kits include an inlet adapter (5.5 to 6.5mm accommodation range); a full-length vacuum adapter with 105° bend; a distilling adapter; an inlet (bleed-in) adapter; a full-length, three-way Claisen adapter; and a 25mL, 50mL, 100mL, 250mL, and 500mL round-bottom flask with a short neck. Each kit also includes a pennyhead stopper, a 125mL funnel, a Liebig condenser, a West condenser, and a storage case.
Customers Who Bought This Also Bought