Synthware Organic Chemistry Kits
Fournisseur: Kemtech America
Certificats
A propos de cet article
For use in college organic laboratory teaching courses.
- Heavy wall borosilicate construction
- Ground glass joints
- 14 Pieces
- 14/20, 19/22, or 24/40
Ordering information: Packaged in a paper carton.
Spécifications
- Catalog No:
- 60004-240
- 60004-242
- Kit contents:
- Claisen Adapter 60000-660, 75° Angle Distillation Adapter 60000-598, Vacuum Take-Off Adapter 60000-636, Inlet/Thermometer Adapter 60000-140, West Condenser with 190 mm Jacket 60000-974, Distillation Column with 200 mm Jacket 60000-800, 25 mL Round-Bottom Flask 60002-288, 50 mL Round-Bottom Flask 60002-294, 100 mL Round-Bottom Flask 60002-300, 250 mL Round-Bottom Flask 60002-314, 500 mL Round-Bottom Flask 60002-328, 125 mL Separatory Funnel 60003-864, Glass Pennyhead Stopper 60004-804
- —