Barnstead/Thermolyne® Thermo Cryogenic Transfer Vessels, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
These dewars and cylinders offer a safe and efficient way to store and dispense small amounts of liquid nitrogen
Lightweight design makes them ideal for everyday laboratory use. Durable construction ensures years of trouble free service even with rough handling associated with portable cryogenic equipment. Withdrawal devices allow easy dispensing of up to 8L/min.
Spécifications
- Catalog No:
- 24269-056
- 24269-054
- 24269-052
- 24269-050
- Model:
- Thermo 30
- Thermo 20
- Thermo 10
- Thermo 5
- Canister height:
- 61 cm (24")
- 63 cm (2413/16")
- 55 cm (2111/16")
- 46 cm (181/8")
- Canister Ø:
- 41 cm (163/16")
- 36 cm (143/16")
- 26 cm (101/4")
- 22 cm (811/16")