Synthware Organic Chemistry Kit, [ST] 14/20, Kemtech America
# de catalogue 60004-238
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Synthware organic chemistry kit, [ST] 14/20
- Kit contents:Claisen adapter 60000-658, 75° angle distillation adapter 60000-596, vacuum take-off adapter 60000-634, inlet/thermometer adapter 60000-138, west condenser with 190 mm jacket 60000-970, distillation column with 200 mm jacket 60000-796, 25 ml round-bottom flask 60002-286, 50 ml round-bottom flask 60002-292, 100 ml round-bottom flask 60002-298, 250 ml round-bottom flask 60002-312, 500 ml round-bottom flask 60002-326, 125 ml separatory funnel 60003-862, glass Pennyhead stopper 60004-802
- Supplier No.:K161420
Spécifications
A propos de cet article
For use in college organic laboratory teaching courses.
- Heavy wall borosilicate construction
- Ground glass joints
- 14 pieces
Ordering information: Packaged in a paper carton.
Customers Who Bought This Also Bought