Cleanroom Wafer Box Tapes, Polyethylene, Ultratape
Fournisseur: Ultratape Industries
Certificats
A propos de cet article
This tape is designed specifically for sealing wafer boxes and cleanroom packaging and bag sealing.
- No residues after prolonged storage
- Easy release from the roll with low static charge
Tape adheres lightly to plastics, stretches easily around corners, and has low metal content.
Delivery: Supplied clean and double-bagged.
Spécifications
- Catalog No:
- 11212-300
- 11212-348
- 11212-302
- 11212-386
- 11212-336
- 11212-390
- 11212-342
- 11212-394
- 11212-338
- 11212-392
- 11212-346
- 11212-344
- 11212-356
- 11212-352
- 89203-784
- 11212-322
- 11212-014
- 11212-306
- 11212-360
- 11212-358
- 11212-310
- 11212-364
- 11212-018
- 11212-022
- 11212-316
- 11212-368
- 11212-020
- 11212-314
- 11212-366
- 11212-372
- 11212-320
- 11212-024
- 11212-318
- 11212-370
- 11212-380
- 11212-376
- Thickness:
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil