Synthware Microscale Kit, Threaded, 14 Pieces, Kemtech America
Catalog # 60004-234
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Synthware Microscale Kit
- Kit contents:Claisen Adapter 60004-112, Distillation Adapter 60004-110, Vacuum Take-Off Adapter 60004-114, Air Reflux Condenser 60004-116, Jacket Condenser 60004-124, Drying Tube 60004-132, Hickman Still Head 60004-134, 2 mL Craig Recrystallization Tube 60004-148, 3 mL Heavy Wall Conical Vial 60004-168, 5 mL Heavy Wall Conical Vial 60004-170, 10 mL Round Bottom Flask 60004-184, 25 mL Round Bottom Flask 60004-186, 10 mL Pear-Shaped Flask 60004-190, PTFE Spinvane 60004-232, Polypropylene Box with Adjustable Compartments 60004-236
- Cat. no.:60004-234
- Supplier No.:K151410
Specifications
About this item
This kit contains 14 components for use in microscale experiments.
- Heavy wall borosilicate construction
- Threaded ground glass joints
- 14 Pieces
Customers Who Bought This Also Bought