Micro Kit for Organic and Environmental Chemistry, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
This kit provides apparatus for a wide range of organic chemistry.
- Smooth surface resist contamination
- Cleaning is simple
- Can be used interchangeably with any standard taper ground joints
This innovative microscale kit offers many advantages over traditional standard glassware, due to the reduced quantity of chemicals required for experiments
Standard taper Clear-Seal® grease-free joints provide smooth, unground surfaces and can be used interchangeably with any standard taper ground joints. All components are manufactured from Wheaton 33* low extractable borosilicate glass (meets ASTM Type I Class A and USP Type I standards).
This results in less chemical waste disposal, less solvent consumption, reduced risk of fire or explosion, the virtual elimination of exposure to toxic materials and vapors, quicker setup times, and shorter reaction times.
Delivery: Complete kit contains 1 Claisen Adapter, 1 Connecting Adapter, 1 Thermometer Adapter, 1 Vacuum Distilling Adapter, 1 West Condenser (200mm), 1 Distilling Column (200mm), 5 Round-Bottom Flasks (25, 50, 100, 250, 500mL), and 1 Separatory Funnel (125mL) with Stopcock. Kit is packaged in a high-density polyethylene case with a foam insert to protect components.
Customers Who Bought This Also Bought