Cleanroom Vinyl Tapes, CR100PC™, Micronova
Supplier: Micronova
Certificates
About this item
Extremely conformable, vinyl cleanroom tape with synthetic rubber, clean-peel adhesive; ideal for colour coding, sealing and marking on curved and irregular surfaces in cleanroom maintenance and support areas.
- Clean peel from wafer cartridges and cleanroom work surfaces
- Synthetic rubber adhesive ensures a clean peel from most cleanroom surfaces such as polyethylene, plastics and stainless steel
- Matte finish can be written on with a cleanroom pen
- Ideal for taping and sealing curved and irregular surfaces
- Special processing and packaging techniques ensure low particles and extractable ion counts for applications in and around the cleanroom
- Wound on a 76 mm (3") plastic core
Length: 33 m (36 yds.)
Available in a variety of colours and widths.
Certifications: Complies with Federal Specification PPP-T-66E and CID A-A 1689A.
Specifications
- Catalog No:
- 68399-226
- 68399-224
- 68399-164
- 68399-162
- 68399-236
- 68399-102
- 68399-234
- 68399-178
- 68399-106
- 76214-310
- 68399-152
- 68399-150
- 68399-215
- 68399-156
- 68399-154
- 68399-194
- 68399-260
- 68399-258
- 68399-200
- 68399-270
- 68399-198
- 68399-140
- 68399-204
- 68399-138
- 68399-202
- 68399-274
- 68399-210
- 68399-250
- 68399-248
- 68399-116
- 68399-188
- Environmentally Preferable:
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient
- Sustainable Packaging, Sustainable Materials, Energy Efficient, Water Efficient