PCX™ Cleanroom Tape, Polyethylene, Micronova
Supplier: Avantor
Certificates
About this item
Low density polyethylene tape, low in halogens and leachable chlorides.
- Special construction lessens the chance of outgassing
- Chemical resistance and good conformability
Tape can be used for color-coding, sealing, labeling and masking. It can be also used for various other tasks within the cleanroom that require pressure-sensitive qualities. Low-density polyethylene tape is low in halogens and leachable chlorides. Packaged in a cleanroom with special processing to ensure low extractable ions, down to parts per billion.
Ordering information: For additional colors and widths, contact your Avantor sales representative.
Customers Who Bought This Also Bought