VWR® Construction Tape, PE
Supplier: VWR International
Certificates
About this item
This high-tack tape has rubber adhesive that easily bonds to plastic, concrete, and stainless steel.
- Excellent conformity
- Serrated edge for easy tearing by hand
- Polyethylene fabrication
Tape conforms well and does not have the same “dead stretch” limitations common in other polyethylene tapes. Unique serrated edge allows an easy tear at any length, for environments where blades and dispensers are not practical.
Customers Who Bought This Also Bought