VWR® Wafer Box Sealing Tape, PE
Fournisseur: VWR International
Certificats
A propos de cet article
Ideal for packaging and transporting of wafer boxes, this low-density polyethylene tape is low in halogens and leachable chlorides.
- Metal free adhesive
- Polyethylene film fabrication
- Acrylic adhesive minimizes outgassing
- Excellent chemical resistance
- Good conformability
Polyethylene film and acrylic adhesive minimize outgassing and offer good 'clean peel' properties. Offers excellent chemical resistance and good conformability.
Spécifications
- Catalog No:
- 89501-298
- 89133-258
- 11211-930
- 11211-928
- 11211-934
- 11211-910
- 11211-932
- 43300-976
- 11211-922
- 89402-996
- 11211-920
- 11211-926
- 11211-924
- Environmentally Preferable:
- Waste Reducing