A propos de cet article
Small, quiet shakers ensure long life with ideal swivel motion.
The basic model has a timer with a range of 5 to 50 minutes and an LED display for speed and time adjustment. The control model features a timer range of 0 to 9 hours, 59 minutes, as well as a digital display with speed, timer function, and operating mode information. The control model also features integrated end point positioning for automated robot-controlled sampling. All functions on the control model can be operated and documented with labworldsoft* software using the RS-232 interface.
Universal platform is for use with various types of vessels. Includes three universal, infinitely variable clamping rolls. Universal platform surface dimensions: 22×22 cm (8¹¹/₁₆×8¹¹/₁₆"). Flask clamp platform is for shaking round flasks, measuring flasks, and Erlenmeyer flasks. Accommodates twenty 25 ml flasks, twelve 50 or 100ml flasks, six 250 ml flasks, or four 500 ml flasks. Flask clamps are available separately. Flask clamp platform surface dimensions: 22.9×22.9 cm (9x9"). Dish platform is suitable for smooth shaking operations in the low viscosity range, such as those involving petri dishes or culture bottles. Includes an integrated slip-resistant foil. Dish platform surface dimensions: 22×34 cm (8¹¹/₁₆×13³/₈"). Test tube rack is for intensive shaking of up to 64 tubes, 10 to 16 mm in diameter and at least 8 cm high. Test tube rack dimensions: 22W×22.9D×9.5H cm (8¹¹/₁₆×9×3³/₄").
Speed and timer are electronically adjustable. A wide range of attachment combinations makes it possible to use almost all shapes and sizes of vessels with these shakers.
Spécifications
- Catalog No:
- 33998-352
- 33998-356
- Max. load:
- 2 kg (4.4 lbs.)
- 2 kg (4.4 lbs.)
- Dimensions:
- 27W x 31.6D x 9.8H cm (105/8 x 127/16 x 37/8")
- 27W x 31.6D x 9.8H cm (105/8 x 127/16 x 37/8")
- Operating temperature range:
- 5...40 °C (41...104 °F)
- 5...40 °C (41...104 °F)