Nalgene® 50 Platinum-Cured Silicone Tubing, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Flexible, durable, high-purity tubing is designed for a variety of pump and transfer applications, including those in the pharmaceutical, research laboratory, bioprocess manufacturing, food and beverage, and semiconductor industries.
- Contains no phthalate plasticizers that can leach out into transported fluid
- Durometer hardness: Shore A, 50
- Autoclavable
Tubing resists adhesion of tissue, blood, proteins, and food products while imparting no taste or odor to the fluids. Manufactured with NALGENE® 50 silicone elastomer. Translucent, nontoxic, nonpyrogenic, and nonhemolytic.
Certifications: Complies with USDA, USP Class VI, and 3A standards.
Ordering information: Supplied in 50' coil lengths.
Caution: Note: Not intended for invasive use.
Specifications
- Catalog No:
- 63014-732
- 63014-734
- 16211-316
- 63014-730
- 16211-320
- 16211-324
- 63014-738
- 16211-340
- 16211-342
- 16211-344
- 16211-326
- 16211-332
- 16211-334
- 16211-338
- Model:
- 63014-732
- 63014-734
- 16211-316
- 63014-730
- 16211-320
- 16211-324
- 63014-738
- 16211-340
- 16211-342
- 16211-344
- 16211-326
- 16211-332
- 16211-334
- 16211-338
- Max. pressure:
- 16 psig at 23°C (73°F), 10 psig at 160°C (320°F)
- 17 psig at 23°C (73°F), 10 psig at 160°C (320°F)
- 15 psig at 23°C (73°F), 9 psig at 160°C (320°F)
- 14 psig at 23°C (73°F), 8 psig at 160°C (320°F)
- 13 psig at 23°C (73°F), 8 psig at 160°C (320°F)
- 12 psig at 23°C (73°F), 7 psig at 160°C (320°F)
- 14 psig at 23°C (73°F), 8 psig at 160°C (320°F)
- 12 psig at 23°C (73°F), 7 psig at 160°C (320°F)
- 12 psig at 23°C (73°F), 7 psig at 160°C (320°F)
- 9 psig at 23°C (73°F), 5 psig at 160°C (320°F)
- 17 psig at 23°C (73°F), 10 psig at 160°C (320°F)
- 11 psig at 23°C (73°F), 7 psig at 160°C (320°F)
- 12 psig at 23°C (73°F), 7 psig at 160°C (320°F)
- 10 psig at 23°C (73°F), 6 psig at 160°C (320°F)