VWR® Latex Rubber Tubing, Black
Fournisseur: VWR International
Certificats
A propos de cet article
Black latex rubber tubing withstands repeated sterilization and has superior tensile strength.
- Durometer hardness: Shore A, 35
- Tensile strength: 3500 psi
- Packaged on 50' reels or in box packs
Black rubber tubing has maximum resistance to aging. Withstands repeated sterilization.
Customers Who Bought This Also Bought