VWR® Three-Sided Enclosure Carts with Round Posts
Supplier: VWR International
Certificates
About this item
Detachable wire mesh panels allow secure handling of objects during transport.
- Secure handling of objects
- Truss-reinforced edges
- Exceptional durability
- Adjustable shelf height
- Round post design
Wire shelves and components feature high-quality, bright chrome plating. Shelves have truss-reinforced edges for exceptional durability. Posts also feature groove rings in 2.5 cm (1") increments for shelf height adjustability. Carts can support up to 544kg (1200lbs.) when weight is evenly distributed.
Ordering information: Supplied with four shelves, four corner posts, four 12.7 cm (5") swivel casters, and detachable panels.
Customers Who Bought This Also Bought