KIMAX® Capillary Tubing, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Tubing is made of KG-33 borosilicate glass, which has a low coefficient of expansion and a high resistance to chemical attack
Ends are cut. Length: 121.9cm ±0.6 (48" ±1/4).
Certifications: Meets requirements for Type I Class A, ASTM E438.
Customers Who Bought This Also Bought