KIMAX® Glass Tubing, Special Wall, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
KG-33 borosilicate glass has a low coefficient of expansion and a high resistance to chemical attack.
Length: 121.9 cm ±0.6 (48" ±¹/₄).
Certifications: Meets requirements for Type I Class A, ASTM E438.
Customers Who Bought This Also Bought