KIMAX® Glass Tubing, Medium Wall, Kimble Chase
# de catalogue 89003-232
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Tubing O.D.:44.5 mm
- Wall thickness:2.4 mm
- Description:Cut Ends
- Cat. no.:89003-232
- Supplier No.:80400-134
Spécifications
A propos de cet article
KIMAX brand KG-33 borosilicate glass has a low coefficient of expansion and a high resistance to chemical attack
Tubing with glazed ends has a length of 121.9 cm ±0.6 (48" ± ¹/₄). Tubing with cut ends has a length of 122.6 cm ±0.6 (48 ¹/₄" ± ¹/₄).
Certifications: Meets the requirements for Type I Class A, ASTM E438.
Customers Who Bought This Also Bought