KIMAX® Glass Tubing, Standard Wall, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
KIMAX® brand KG-33 borosilicate glass has a low coefficient of expansion and a high resistance to chemical attack.
Tubing with glazed ends has a length of 121.9 cm ±0.6 (48" ±¹/₄). Tubing with cut ends has a length of 122.6 cm ±0.6 (48¹/₄" ±¹/₄).
Certifications: Meets the requirements for Type I Class A, ASTM E438.
Customers Who Bought This Also Bought