Spectra/Por® Weighted Closures, Spectrum® Laboratories
Supplier: Spectrum Laboratories
Certificates
About this item
Pressure-fitted with an acid-washed, stainless steel bar into the reverse side
Made of polypropylene. Autoclavable. When used in tandem with the standard closure, weighted closure facilitates complete submersion of the dialysis sac for a more efficient dialysis effect.
Customers Who Bought This Also Bought