Impact Mortar and Pestle, Chemplex®
# de catalogue 15566-088
Fournisseur: Chemplex
Spécifications
- O.D.:9 cm (3.5")
- Volume:70 ml
- Height:19 cm (7.52)
- Supplier No.:850
Spécifications
A propos de cet article
Crushes and pulverizes rocks and similar sample materials for spectrochemical analysis
Unit consists of a cylindrical pestle and a mortar sample chamber. All components disassemble for cleaning.
Customers Who Bought This Also Bought