Spécifications
- Description:Surgipath® Cassette Storage Drawers
- Cat. no.:95057-864
- Supplier No.:39MC-900-1
Spécifications
A propos de cet article
Drawers offer an efficient means of organizing and filing histology case blocks for future reference.
- Efficient organization
- High polymer resin
- Houses 256 cassettes
Drawers offer an efficient means of organizing and filing histology case blocks for future reference.
Constructed of a high polymer plastic resin, the storage system houses 256 standard cassettes and approximately 500 mini cassettes. Each two drawer system can be used individually for transport, or combined with additional drawers and stacked to create a larger system.