Hydraulic Pedalifts, Wesco
Fournisseur: Wesco
Certificats
A propos de cet article
Rugged, steel frame pedalifts provide a lift capacity of 340kg (750lbs.)
Pedalift 21002-042 features two 20.3x5cm (8x2") wheels with 1.9cm (3/4") ball bearings. The front tip of the nose plate touches the floor.
Pedalift 21002-044 features two 20.3x5cm (8x2") wheels with 1.9cm (3/4") ball bearings and two 8.8cm (31/2") diameter polyurethane ball bearing front swivel casters. The platform sits 14.6cm (53/4") above the floor.
Wheel locks are standard on both models.
Foot-operated hydraulic pump raises platform; foot release lowers platform. Lift height: 137.1cm (54"). Overall height: 153.6cm (601/2"). Platform/fork dimensions: 55.8Dx55.8Wcm (22x22"). Available with two or four wheels.
Customers Who Bought This Also Bought