Nalgene® 489 Low-Density Polyethylene Tubing, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Translucent polyethylene tubing for laboratory and industrial use.
Contains no plasticizers. Nontoxic, odorless, and tasteless. Superior chemical resistance. Not autoclavable, but may be chemically sterilized. Durometer hardness: Shore D, 50. Complies with FDA Regulations for food use.
Ordering information: Supplied in 50' or 100' coil lengths.
Specifications
- Catalog No:
- 63006-089
- 63006-103
- 63006-045
- 63006-067
- 63006-012
- Model:
- 63006-089
- 63006-103
- 63006-045
- 63006-067
- 63006-012
- Max. pressure:
- 118 psig @ 23 °C (73 °F), 59 psig @ 63 °C (145 °F)
- 96 psig @ 23 °C (73 °F), 48 psig @ 63 °C (145 °F)
- 214 psig @ 23 °C (73 °F), 107 psig @ 63 °C (145 °F)
- 152 psig @ 23 °C (73 °F), 76 psig @ 63 °C (145 °F)
- 358 psig @ 23 °C (73 °F), 179 psig @ 63 °C (145 °F)