Chemfluor® Fluoropolymer Tubing, Formulation 367, Saint-Gobain Life Sciences
Supplier: Saint Gobain Performance Plastics
Certificates
About this item
Ideal for laboratory and pharmaceutical use, or for applications where the process is dependent on maintaining high purity and fluid integrity.
- Fluoropolymer tubing outperforms tubing made from traditional or high-purity PFA resins
- Features very low extractables with deionized water and acids
Superior clarity and surface smoothness reduce the risk of cross-contamination, improve product yields, and help ensure thorough cleaning cycles. Resistant to virtually all chemicals.
Temperature range: –240 to +232 °C (–400 to +450 °F). Durometer hardness: Shore D, 58. Tensile strength: 2300psi. Autoclavable. Available in 15.2 m (50') coil lengths.
Customers Who Bought This Also Bought