Grade A™ Food Production Mats, Wearwell®
Fournisseur: Wearwell
Certificats
A propos de cet article
Designed specifically for the food production industry
Specially formulated antimicrobial nitrile rubber composite will withstand solutions used in a wide variety of food processing environments: vegetable oils, animal fats, citrus, and fish oil. The built-in handles allow for easy handling and cleaning. The hygienic porthole and solid underside design simplify cleaning and virtually eliminate areas where food and bacteria can collect. The 6mm (1/4") high underside knobs increase comfort level, while the flat textured surface increases traction when greasy and wet. Thickness: 1.3cm (1/2").
Customers Who Bought This Also Bought