Versilon™ Fluoropolymer Tubing, PTFE, FEP, or PFA
Fournisseur: Saint Gobain Performance Plastics
Certificats
A propos de cet article
Fluoropolymer tubing is recommended in many demanding areas such as chromatography, semiconductor processing, Type I Water, and cryogenics.
- Tubing offers the ultimate in purity
- Chemical resistant
- Broad temperature capability for handling ultrapure fluids or highly corrosive materials
Choose from PTFE, FEP, or PFA fluoropolymer resin. Contains no additives or plasticizers to contaminate liquids. Inert to virtually all chemicals. Not for use in peristaltic pumps.
PTFE tubing has the broadest temperature range: –268 to +288 °C (–450 to +550 °F). Fully-fluorinated with excellent chemical inertness. FDA compliant.
FEP tubing has the greatest clarity and abrasion resistance. Transparent to UV light. FDA compliant.
PFA tubing has very low metal ion content for trace metal analysis and superior strength and burst resistance at elevated temperatures.
Ordering information: Supplied in 50' coil lengths.
Spécifications
- Catalog No:
- 63014-861
- 63014-960
- 63014-961
- 63014-934
- 63014-690
- 63014-860
- 63014-950
- 63014-915
- 63014-815
- 63014-925
- 63014-809
- 63014-914
- 63014-699
- 63014-696
- 63014-800
- 63014-905
- 63014-805
- 63014-695
- Model:
- 63014-861
- 63014-960
- 63014-961
- 63014-934
- 63014-690
- 63014-860
- 63014-950
- 63014-915
- 63014-815
- 63014-925
- 63014-809
- 63014-914
- 63014-699
- 63014-696
- 63014-800
- 63014-905
- 63014-805
- 63014-695
- Max. pressure:
- 65 psig @ 23°C (73°F)
- 95 psig @ 23°C (73°F)
- 65 psig @ 23°C (73°F)
- 120 psig @ 23°C (73°F)
- 440 psig @ 23°C (73°F)
- 95 psig @ 23°C (73°F)
- 270 psig @ 23°C (73°F)
- 230 psig @ 23°C (73°F)
- 230 psig @ 23°C (73°F)
- 155 psig @ 23°C (73°F)
- 155 psig @ 23°C (73°F)
- 115 psig @ 23°C (73°F)
- 155 psig @ 23°C (73°F)
- 230 psig @ 23°C (73°F)
- 440 psig @ 23°C (73°F)
- 455 psig @ 23°C (73°F)
- 455 psig @ 23°C (73°F)
- 115 psig @ 23°C (73°F)