24/Seven™ Modular Floor Mats, Wearwell®
Fournisseur: Wearwell
Certificats
A propos de cet article
These 0.9x0.9m (3x3') modular, molded rubber antifatigue mats are resilient and comfortable.The mats are available with a functional and effective drainage surface, or with a solid surface for use in mostly dry areas. Both styles are available in three unique compounds to address the diverse environments found in typical work applications.
- The grease-resistant mats are ideal for general-purpose applications
- The cutting-fluid-resistant (CFR) compound mats are ideal for areas with mineral oil-based cutting fluids
- The 100% nitrile compound mats are ideal for areas with petroleum-based fluids, such as hydrocarbons, naphthenes, paraffins, petroleum distillates, and other petrochemicals
- For additional traction GritWorks!® surface on the CFR or nitrile mats: provides up to 40% more traction than recommended by OSHA in wet areas
- Single workstation mats also have a hidden interlocking system designed to connect on all four sides to quickly cover large areas
For extra safety, attach optional yellow 7.6Wx99Lcm (3x39") beveled ramp edges. Female ramps snap onto the protruding mat knobs; male ramps snap into the underside of the mat.
All mats are 1.6 cm (⅝") thick.
Customers Who Bought This Also Bought