ContactUsLinkComponent
Blade Safebox, Martor USA
Blade Safebox, Martor USA
# de catalogue 89037-522
Fournisseur:  Martor
Used Blade Container
Blade Safebox, Martor USA
# de catalogue 89037-522
Fournisseur:  Martor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Blade Safebox, plastic
  • Cat. no.:
    89037-522
  • Supplier No.:
    108000
Spécifications
A propos de cet article
The SAFEBOX is large enough to store a large number of used blades safely. And at the same time, it is compact enough to carry with you.
The two slits are not only used for filling, but also to "shorten" your snap-off blades. When your Safebox is full, empty it into a container for used blades.
Customers Who Bought This Also Bought