Accessories for UVP High-Intensity UV Inspection Lamps, Analytik Jena
Fournisseur: Analytik Jena US
Certificats
Certificats
A propos de cet article
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Fournisseur: VWR International
$109.33 to $364.87
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Fournisseur: MilliporeSigma
$1,313.27 to $5,149.78
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$194.14 to $694.12