SP Bel-Art Safety Step Stool, Bel-Art Products, a part of SP
Catalog # 59075-022
Supplier: Bel-Art Products, a Part of SP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Load Capacity:330 lbs.
- Description:Safety step stool
- Height:12"
- Cat. no.:59075-022
- Supplier No.:249000000
Specifications
About this item
This handy step stool provides safe, easy access to high shelves, drawers, equipment and other hard-to-reach areas.
- Three spring-loaded, swiveling casters make it easy to kick or push out of the way or into position
- When weight is placed on the stool a non-skid vinyl bumper rests directly on the floor to prevent movement
- Easy to clean
Base diameter: 15⁵/₈"
Shipping weight: 7 lbs.
Certifications: EN 141183
Customers Who Bought This Also Bought