VWR® Talon® Support Plates
Supplier: VWR International
Certificates
About this item
Designed to hold hotplate/stirrers and other apparatus to lab frames or ring stands.
- Aluminum construction
- Strong and durable
- Now with Best-in-Class, Exclusive five year warranty
Plates include holder for mounting to vertical rod up to 1.9 cm (³/₄") and a self-adhesive non-skid mat
Use the small support plate for most 10.2×10.2 cm (4×4") hotplate/stirrers; the medium for 17.8×17.8 cm (7×7") hotplate/stirrers; and the large for multiple units.
Aluminum construction offers strength and durability.
Specifications
- Catalog No:
- 11301-126
- 11301-124
- 11301-128
- Environmentally Preferable:
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging