Stainless Steel Step Stool, Bandy
Catalog # 24450-004
Supplier: Bandy
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Width:24"
- Description:Stainless steel 2-step stool
- Height:54"
- Length:30"
- Cat. no.:24450-004
- Supplier No.:STP00001
Specifications
About this item
Lightweight, two-level step stool for the cleanroom
Can be hand carried. Constructed of 16-gauge, type 304 stainless steel and completely electropolished. Adjustable leveling feet.
Customers Who Bought This Also Bought