Spécifications
- Volume:100 µl
- Description:SureSTART™ WebSeal™ 96-well microtiter plates, total V-bottom, Level 2 high-throughput applications
- Material:Polypropylene
- I.D.×H:5.6×15 mm
- Well Shape:Conical Bottom (V-well)
Spécifications
A propos de cet article
SureSTART™ Level 2 WebSeal™ 96-Well Total V-Bottom microtiter plates are designed to ensure high quality data with an uninterrupted workflow for high-throughput GC and HPLC/UHPLC applications.
- Level 2 High-throughput applications
Ideal for non-regulated labs that use water and polar solvents
Certified plastic well plates, seals, tapes
GC-MS and LC-MS tested
Suitable for HPLC/UHPLC applications
Suitable for QA/QC workflows and sensitive applications with low sample concentrations
Ideal for verifying compound presence and quantification using detection techniques such as UV, DAD/PDA, RI, Fluorescence, and ELSD
Specification Certified
SureSTART WebSeal well plates are dimensionally verified
They are ideal for regulated labs that use water and polar solvents. Use Level 2 polypropylene well plates for routine QA/QC workflows, sensitive applications with lower sample concentrations, and data collection for total ion chromatograms (TIC). These 96-well plates are certified to ensure autosampler compatible physical dimensions and conform to ANSI-SLAS 4-2004 standards.