SureSTART™ WebSeal™ 96-Well Mid-well Plates, Square U-Bottom
# de catalogue 75982-330
Fournisseur: Thermo Scientific
Spécifications
- Volume:1600 µl
- Description:SureSTART™ WebSeal™ 96-Well mid-well plates, square U-bottom, clear, Level 1 everyday analysis
- Height:31 mm
- Material:Polypropylene
- Well Shape:Square, round bottom (U-well)
Spécifications
A propos de cet article
SureSTART™ Level 1 WebSeal™ 96-Well Square U-Bottom Mid-well plates are designed for everyday chromatography analysis.
- Ideal for non-regulated labs that use water and polar solvents
- Commodity plates, seals, tapes
- GC tested
- Suitable for HPLC/UHPLC applications
- Suitable for biopharmaceutical applications
- Suitable for QA/QC applications using core detectors
- Ideal for verifying compound presence and quantification using detection techniques such as UV, DAD/PDA, RI, Fluorescence, and ELSD
- SureSTART WebSeal well plates are dimensionally verified
- Non sterile and non coated
- 8×12 array
They provide an affordable choice for use with most HPLC/UHPLC instrument types, and are ideal for non-regulated laboratories using water, water and polar solvents and polar solvents. These 96-well plates are specification certified to ensure autosampler compatible physical dimensions and conform to ANSI-SLAS 4-2004 standards.
Certifications: GC/MS Tested.
Customers Who Bought This Also Bought