Spectrocertified® Pre-Fused Fusion Fluxes, Chemplex®
Fournisseur: Chemplex
Certificats
A propos de cet article
Pre-fused fusion fluxes are for preparing powdered samples for XRF, ICP, and AA analyses
- Fluxes feature <0.03% loss on ignition and low eutectic melting points for effective dissolution. High density prevents spillover in crucibles with low volumetric capacity. Anhydrous nature reduces water content and eliminates drying. Granulation promotes uniform particle size. Less than 0.05% of particles in mixture are <100 µm. Free-flowing mixtures may be handled manually or through a dispenser. Technical specifications included on bottle label.
Packaging: Packaged 454 g (1 lbs.) per bottle.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.