ContactUsLinkComponent
Corning® PC-220 Low Profile Hotplate Stirrer, Corning
Corning® PC-220 Low Profile Hotplate Stirrer, Corning
# de catalogue 89090-994
Fournisseur:  Corning
cUL Listed
CSA Certified
Product available on GSA Advantage®
UL Listed
HOT PLATE/STIRRER PC220 4X5 120 V CS1
Corning® PC-220 Low Profile Hotplate Stirrer, Corning
# de catalogue 89090-994
Fournisseur:  Corning
cUL Listed
CSA Certified
Product available on GSA Advantage®
UL Listed
Spécifications
  • Temperature Range:
    25…550 °C (77…1022 °F)
  • Description:
    PC-220
  • Electrical:
    120 V, 60 Hz
  • Speed Range:
    60 - 1150 rpm
  • Top Plate Dimensions:
    4×5"
  • Supplier No.:
    6795-220
Spécifications
A propos de cet article
Superior performance and reliability in a low profile design.
The Pyroceram® glass ceramic top heats evenly, is highly resistant to scratches, corrosion, and chemical attack, and wipes clean with a damp cloth. A rugged, double ball bearing motor and other high quality components prolong product life.
  • Micro-chemistry size
  • Hot top indicator light
  • Glass ceramic top
  • Two-year warranty
This Corning® hotplate stirrer has a durable glass-ceramid 4×5" Pyroceram® top and offer excellent performance and reliability in a space-saving design. It offer closed-loop stirring for consistent and accurate speed and to get the job done quickly, efficiently, and safely. A hot top indicator light blinks a warning if the top is too hot to touch (>140 °F) - even when the unit is turned off. A built in holder accommodates optional 4" stainless steel support rods. 120 V units are available and are UL and CUL approved to meet needs in North America and other locations around the world. A 1׳/₈" stir bar is included with each stirrer.
Certifications: UL and C-UL listed. CE marked. FCC, IEC1010, and GS certified.
Ordering information: Supplied with manufacturer's two-year warranty on parts and labor. Includes stir bar and instructions.
Customers Who Bought This Also Bought