XRF Sample Cup Series 1900, Chemplex®
Fournisseur: Chemplex
Certificats
A propos de cet article
Polyethylene sample cups intended for handling large numbers of high density loose powder and non-volatile liquid samples in air or inert gas environments
Consists of a double open-ended cell, snap-on ring and collar. Cell neck and snap-on ring incorporate a tapered “bead-to-indent” geometry for clamping thin-film sample supports. Collar is used to temporarily secure a thin-film support to the cell followed by attachment with a snap-on ring. 15566-172 is for use with Oxford Lab-X analytical systems.
Customers Who Bought This Also Bought