XRF Sample Cup Series 1000, Chemplex®
Fournisseur: Chemplex
Certificats
A propos de cet article
Polyethylene sample cups are designed to eliminate trimming of extraneous thin-film sample supports and facilitate general sample preparation procedure.
A full length friction-fitting sleeve secures and envelopes the thin-film sample support to the cell body. Available in single or double open-ended versions. Single open-ended sample cups have an overflow reservoir and a thermoplastic seal that can be optionally ruptured to equalize pressure. Double open-ended cups permit attachment of thin-film sample supports prior to sample introduction for applications in air or inert gas environments. Height: 25mm (1").
Customers Who Bought This Also Bought