A propos de cet article
The PC-410D stirrers have a Pyroceram® top and digital stirring speed display.
All units have a small footprint and low profile to ensure a comfortable fit inside hoods or storage cabinets. An automatic ramp-up speed adjustment feature minimizes decoupling. The extremely durable and heat-resistant Pyroceram tops are easy to clean and highly resistant to scratches, corrosion, and chemicals. The angled front panel and large knob design minimize damage resulting from spills.
- Closed-loop stirring control monitors and regulates the stirring speed for aqueous, viscous, or semi-solid solutions
- Stronger, larger magnet improves stir bar coupling combined with automatic ramp up speed adjustment which minimises decoupling
- Design deflects spills away from electronics and includes a built in mount for an optional support rod
The PC-410D stirrers have a Pyroceram® top and digital stirring speed display.
All units have a small footprint and low profile to ensure a comfortable fit inside hoods or storage cabinets. The extremely durable and heat-resistant Pyroceram tops are easy to clean and highly resistant to scratches, corrosion, and chemicals. The angled front panel and large knob design minimize damage resulting from spills.
All stirrers feature Pyroceram® glass-ceramic tops set atop rugged housing. Digital stirrers have a closed-loop stirring control that monitors and regulates speed.
Spécifications
- Catalog No:
- 89090-244
- 89090-236
- 89090-238
- 89090-246
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa
- I-RECs, ISO 14001 - Reynosa