Bard-Parker® Stainless Steel Scalpel Blades, Nos. 10–15, Sterile, Aspen Surgical
Supplier: Aspen Surgical Products
Certificates
About this item
Stainless steel blades are sterile and individually packaged
For use with No. 3, No. 3L, No. 3LA, No. 5, No. 7, and No. 9 Bard-Parker® nonsterile surgical handles.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.