Chem-Wik® Rosin Desoldering Braids, ITW Chemtronics®
Fournisseur: ITW Chemtronics
Certificats
A propos de cet article
These ultrapure, oxygen-free copper braids quickly and completely remove solder from circuit boards and components
The fast wicking action protects components from heat damage. Braids are coated with ultra-high purity, noncorrosive Type R rosin flux type. Meet DOD-STD-1686, DOD-HDBK-263.
Customers Who Bought This Also Bought