Classic Stills, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Barnstead Classic Stills effectively remove inorganic solids, organics, bacteria, and pyrogens
The stills are constructed of copper and bronze with a coating of pure tin. The inert nature of tin prevents any leaching of contaminants into the water.
Certifications: UL.
Ordering information: Requires storage reservoir, UV light assembly and Ventgard Air Filter (all ordered separately). Fully Automatic Controls (FAC) are available for all Classic Stills (order separately). The FAC system turns the still on/off to ensure a readily available supply of distilled water at all times. The FAC includes feed water and drain solenoid valves, manual valves, piping and level monitors. For more information contact your VWR representative.
Delivery: Includes floor stand (except for portable model A1007).
Customers Who Bought This Also Bought