Barnstead Mega-Pure™ Glass Stills, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Barnstead Mega-Pure Glass Stills effectively remove inorganic solids, organics, with boiling points higher than water, bacteria, and pyrogens
The stills are constructed of non-leaching components to ensure high purity water.
Certifications: CSA.
Ordering information: Automatic Collection System (ACS) is available for all Mega-Pure systems. The ACS automatically turns on/off the still to ensure a ready supply of distilled water at all times. The ACS comes standard with 45L glass storage bottle, float, and controller. For more information contact your Avantor representative.
Customers Who Bought This Also Bought