Stainless Steel Frits, Upchurch Scientific®
Fournisseur: Upchurch Scientific
Certificats
A propos de cet article
These 316 stainless steel frits are the most common size filters used throughout HPLC systems.
Frits are encased in a polymer ring for best performance and proper sealing.
Customers Who Bought This Also Bought